Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse C1qTNF10/C1qL2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7066062
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4374SP 25 μg
MAB4374 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4374SP Supplier R&D Systems Supplier No. MAB4374SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

C1qTNF10/C1qL2 Monoclonal specifically detects C1qTNF10/C1qL2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen C1qTNF10/C1qL2
Applications Western Blot
Classification Monoclonal
Clone 384921
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8CFR0
Gene Alias C1q and tumor necrosis factor related protein 10, C1q-domain containing protein, C1qL2, C1QTNF10, complement C1q-like protein 2, complement component 1, q subcomponent-like 2, CTRP10
Gene Symbols C1QL2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse C1qTNF10/C1qL2 His22-Asp287 Accession # Q8CFR0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165257
Test Specificity Detects mouse C1qTNF10/C1qL2 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant human (rh) C1qTNF10 and no cross-reactivity with rhC1qTNF1, rhC1qTNF4, or rhC1qTNF5.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.