Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mouse C4b Partial ORF (NP_033910.2, 20 a.a. - 119 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89952801 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-952-801 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-952-801 Supplier Abnova™ Supplier No. P3687
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Sequence: KPRLLLFSPSVVNLGTPLSVGVQLLDAPPGQEVKGSVFLRNPKGGSCSPKKDFKLSSGDDFVLLSLEVPLEDVRSCGLFDLRRAPHIQLVAQSPWLRNTA

Specifications

Accession Number NP_033910.2
For Use With (Application) Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 12268
Molecular Weight (g/mol) 36.63kDa
Name Mouse C4b Partial ORF (NP_033910.2, 20 a.a. - 119 a.a.) Recombinant Protein with GST-tag at N-terminal
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at −80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias C4, Ss
Common Name MOUSEC4B
Gene Symbol C4b
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST Tag
Expression System wheat germ expression system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.