Learn More
Sino Biological Mouse CAMK4 MAb 100UL 50838-R001-100
Supplier:  Sino Biological 50838R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CaM Kinase 4 / CaMKIV (rM CaM Kinase 4 / CaMKIV; Catalog#50838-M20B; P08414; Met1-Tyr469).
Catalog No. 50-240-9743
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.