Learn More
Abcam Mouse Cathelicidin/CLP Peptide, Synthetic, >70% Purity, 1 mg
Description
- Target Protein: Mouse Cathelicidin/CLP peptide
- Product Type: Synthetic blocking peptide
- Purity: >70%
- Tested Applications: BL
- Form: Lyophilized
- Storage Conditions: Store at -20°C
- Shipping Temperature: Blue Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Blocker and Diluent |
| Content And Storage | Store at -20°C. |
| Form | Lyophilized |
| Concentration | 1 mg/mL |
| Substrate Properties | Peptide-based Substrate |
| Target Molecule | Cathelicidin/CLP (Mouse) |
| Label or Dye | Unconjugated |
| Reactivity | Mouse |
| Shipping Condition | Blue Ice |
| Cross Reactivity | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.