Learn More
Eurofins Discoverx Recombinant Mouse CCL1/I-309/TCA-3, 25 µg. (92-1199)
Supplier:  Eurofins Discoverx 921199
Encompass
Murine CCL1 is a cytokine protein secreted by activated T cells to attract monocytes, NK cells, and immature B cells by binding with CCR8 cell surface receptors. CCL1 is related to demyelination and may be associated with neuroinflammatory disorders. Murine CCL1 is cross-species reactive with human receptors. Provided as 25 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9727
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.