Learn More
R&D Systems™ Mouse CCL11/Eotaxin Quantikine ELISA Kit
Description
Specifications
Specifications
| Assay Range | 15.60 - 1000pg/mL |
| Assay Sensitivity | 3pg/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate), Serum |
| Specificity | Natural and Recombinant Mouse Eotaxin. This assay cross-reacts 0.062% with Human Eotaxin and recognizes Rat Eotaxin. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 20292 |
| Quantity | 2 x 96-well Plates |
| Sample Volume | Cell Culture Supernates (50uL),Serum (13uL),EDTA Plasma (13uL),Citrate Plasma (13uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.