Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse CCL12/MCP-5 Quantikine ELISA Kit
SDP

Catalog No. MCC120 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MCC120 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MCC120 Supplier R&D Systems™ Supplier No. MCC120

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse CCL12/MCP-5 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse CCL12/MCP-5 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse CCL12/MCP-5 in cell culture supernates (50 ul), serum (25 ul), edta plasma (25 ul).

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 2.1pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA), Serum
Specificity Natural and Recombinant Mouse MCP-5
For Use With (Application) ELISA
Gene ID (Entrez) 20293
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Serum (25uL),EDTA Plasma (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym C-C motif chemokine 12, chemokine (C-C motif) ligand 12, MCP5, MCP-5, Scya12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.