Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse CCL2/JE/MCP-1 Quantikine ELISA Kit
SDP

Catalog No. MJE00B Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MJE00B 1 Kit
SMJE00B 1 Pack (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MJE00B Supplier R&D Systems™ Supplier No. MJE00B

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse CCL2/JE/MCP-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse CCL2/JE/MCP-1 Quantikine ELISA Kit quantifies mouse CCL2/JE/MCP-1 in cell culture supernates (50 ul), tissue lysates (50 ul), tissue culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).

1 Kit

Specifications

Assay Range 7.81 - 500 pg/mL
Assay Sensitivity 0.666 pg/mL
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Lysates
Specificity Natural and Recombinant Mouse MCP-1
For Use With (Application) ELISA
Gene ID (Entrez) 6347
Gene Symbol CCL2
Host Species Mouse
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50 uL),Tissue Lysates (50 uL),Tissue Culture Supernates (50 uL),Serum (50 uL),EDTA Plasma (50 uL),Heparin Plasma (50 uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target CCL2/JE/MCP-1
Total Assay Time 4.5 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.