Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse CCL20 (MIP-3 alpha) Recombinant Protein

Catalog No. PIRP8673
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP8673 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP8673 Supplier Invitrogen™ Supplier No. RP8673

Recombinant Protein

CCL20 (mouse) Recombinant Protein, RP-8673, from Invitrogen.

Macrophage Inflammatory Protein-3 alpha (MIP-3alpha), also called CCL20, is expressed in the liver, lungs, lymph nodes and peripheral blood lymphocytes. MIP-3alpha is strongly up regulated by inflammatory signals, and down regulated by the anti-inflammatory cytokine IL-10. MIP3alpha signals through the G protein-coupled receptor, CCR6, and acts as a chemoattractant to lymphocytes and dendritic cells. Recombinant mouse VEGF-120 is an alternative spliced version of mouse VEGF-165. It is a non-glycosylated homodimer, containing two chains, with a molecular weight of 28.2 kDa. Purity is typically greater than 95% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the dose-dependent proliferation of human umbilical vein endothelial cells (HUVEC) and is typically 1-5 ng/mL. The specific activity of this protein is 1x10^6 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: APTTEGEQKS HEVIKFMDVY QRSYCRPIET LVDIFQEYPD EIEYIFKPSC VPLMRCAGCC NDEALECVPT SESNITMQIM RIKPHQSQHI GEMSFLQHSR CECRPKKDRT KPEKCDKRPRR.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O89093
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 0.1%TFA with no preservative
Gene ID (Entrez) 20297
Molecular Weight (g/mol) 7.9
Name Mouse MIP-3 alpha
Quantity 20 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias M-MIP-3-alpha
Common Name Mouse MIP-3 alpha
Gene Symbol Ccl20
Specific Reactivity Mouse
Conjugate Unconjugated
Cross Reactivity Ms
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitroresearch only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.