Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse CCL20 (MIP-3 alpha) Recombinant Protein, PeproTech®

Catalog No. 25027100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25027100UG 100 μg
250-27-1MG 1 mg
250-27-20UG 20 μg
25027250UG 250 μg
25027500UG 500 μg
250-27-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 25027100UG Supplier Gibco™ Supplier No. 25027100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine MIP-3alpha is a 7.9 kDa protein containing 70 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Macrophage Inflammatory Protein-3 alpha (MIP-3alpha), also called CCL20, is expressed in the liver, lungs, lymph nodes and peripheral blood lymphocytes. MIP-3alpha is strongly up regulated by inflammatory signals, and down regulated by the anti-inflammatory cytokine IL-10. MIP3alpha signals through the G protein-coupled receptor, CCR6, and acts as a chemoattractant to lymphocytes and dendritic cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O89093
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 20297
Molecular Weight (g/mol) 7.9 kDa
Name Mouse CCL20 (MIP-3 alpha)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias beta chemokine exodus-1; beta-chemokine exodus-1; CC chemokine LARC; CC chemokine ST38; C-C motif chemokine 20; C-C motif chemokine ligand 20; Ccl20; CCL20(1-64); CCL20(1-67); CCL20(2-70); chemokine (C-C motif) ligand 20; CKb4; Exodus; exodus-1; LARC; Liver and activation-regulated chemokine; macrophage inflammatory protein 3 alpha; MIP 3 A; MIP 3 A; MIP 3 alpha; MIP 3 A; MIP3 a; MIP3 alpha; MIP3 a; MIP-3 [a]; MIP3A; MIP-3 A; MIP-3-alpha; M-MIP-3-alpha; SCYA20; small inducible cytokine subfamily A (Cys-Cys), member 20; small inducible cytokine subfamily A20; small-inducible cytokine A20; St38
Common Name CCL20
Gene Symbol CCL20
Biological Activity Determined by its ability to chemoattract murine CCR6-transfected HEK293 cells using a concentration range of 0.1-10.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence ASNYDCCLSY IQTPLPSRAI VGFTRQMADE ACDINAIIFH TKKRKSVCAD PKQNWVKRAV NLLSLRVKKM
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.