Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse CCL21 (6Ckine) Recombinant Protein, PeproTech®

Catalog No. 25013100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25013100UG 100 μg
25-013-01M 2 x 500 μg
250-13-20UG 20 μg
25013250UG 250 μg
25013500UG 500 μg
250-13-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 25013100UG Supplier Gibco™ Supplier No. 25013100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

250-13-1MG will be provided as 2 x 500 μg (250-13-500UG). Recombinant Murine Exodus-2 is a 12.0 kDa protein containing 110 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. Similar to other chemokines the protein encoded by this gene inhibits hemopoiesis and stimulates chemotaxis. This protein is chemotactic in vitro for thymocytes and activated T cells, but not for B cells, macrophages, or neutrophils. The cytokine encoded by this gene may also play a role in mediating homing of lymphocytes to secondary lymphoid organs. It is a high affinity functional ligand for chemokine receptor 7 (CCR7) that is expressed on T and B lymphocytes and a known receptor for another member of the cytokine family (small inducible cytokine A19).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P84444, P86792, P86793
For Use With (Application) Control, Western Blot Control, ELISA Standard, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 100042493, 18829, 65956
Molecular Weight (g/mol) 12 kDa
Name Mouse CCL21 (6Ckine)
Purification Method Purified
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 6 Ckine; 6CKBAC1; 6CKBAC2; 6Ckine; AI159701; ALP; AW987545; beta chemokine exodus-2; beta-chemokine exodus-2; CC chemokine 6Ckine-ser; C-C motif chemokine 21; C-C motif chemokine 21a; C-C motif chemokine 21b; C-C motif chemokine 21c; C-C motif chemokine ligand 21; CCL21; Ccl21a; Ccl21b; Ccl21c; Ccl21-leu; CCL21-Ser; chemokine (C-C motif) ligand 21; chemokine (C-C motif) ligand 21A (serine); chemokine (C-C motif) ligand 21B (leucine); chemokine (C-C motif) ligand 21b (serine); chemokine (C-C motif) ligand 21C (leucine); chemokine CCL21/6CKINE; CKb9; ECL; Efficient Chemoattractant for Lymphocytes; exodus-2; plt; Scya21; SCYA21a; Scya21b; Scya21c; secondary lymphoid organ chemokine; secondary lymphoid tissue chemokine; Secondary lymphoid-tissue chemokine; SLC; small inducible cytokine A21; small inducible cytokine A21a; small inducible cytokine A21c; small inducible cytokine subfamily A (Cys-Cys), member 21; small-inducible cytokine A21; Small-inducible cytokine A21a; Small-inducible cytokine A21b; Small-inducible cytokine A21c; TCA4; Thymus-derived chemotactic agent 4; UNQ784/PRO1600
Common Name CCL21
Gene Symbol Ccl21a, Ccl21b, Ccl21c
Biological Activity Determined by its ability to chemoattract human activated T cells cultured in the presence of IL-2 using a concentration range of 10.0-100.0 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence SDGGGQDCCL KYSQKKIPYS IVRGYRKQEP SLGCPIPAIL FLPRKHSKPE LCANPEEGWV QNLMRRLDQP PAPGKQSPGC RKNRGTSKSG KKGKGSKGCK RTEQTQPSRG
Content And Storage -20°C
Activity Determined by its ability to chemoattract human activated T cells cultured in the presence of IL-2 using a concentration range of 10.0-100.0 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.