Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse CCL3 (MIP-1 alpha) Recombinant Protein, PeproTech®

Catalog No. 2500950UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
250-09-50UG 50 μg
25009100UG 100 μg
250-09-10UG 10 μg
250-09-1MG 1 mg
25009250UG 250 μg
250-09-2UG 2 μg
25009500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 250-09-50UG Supplier Gibco™ Supplier No. 2500950UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine MIP-1alpha is a 7.8 kDa protein containing 69 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Both MIP-1alpha and MIP-1beta are structurally and functionally related CC chemokines. They participate in host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells (e.g. macrophages, lymphocytes and NK cells). While both MIP-1alpha and MIP-1beta exert similar effects on monocytes, their effect on lymphocytes differ; with MIP-1alpha selectively attracting CD8+ lymphocytes, and MIP-1beta selectively attracting CD4+ lymphocytes. Additionally, MIP-1alpha and MIP-1beta have also been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. Both human and murine MIP-1alpha and MIP-1beta are active on human and murine hematopoietic cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10855
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 20302
Molecular Weight (g/mol) 7.8 kDa
Name Mouse CCL3 (MIP-1 alpha)
Quantity 50 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AI323804; C-C motif chemokine 3; C-C motif chemokine ligand 3; CCL3; chemokine (C-C motif) ligand 3; G0/G1 switch regulatory protein 19-1; G0S19-1; Heparin-binding chemotaxis protein; H-MIP-1-alpha; L2G25B; LD78ALPHA; LD78-alpha; LD78-alpha(4-69); Macrophage inflammatory protein 1 alpha (Small inducible cytokine A3); macrophage inflammatory protein 1-alpha; macrophage inflammatory protein-1 alpha; MIP1 (a); mip1 alpha; MIP-1 alpha; MIP1-(a); MIP1A; MIP-1 A; MIP-1 alpha; MIP-1-alpha; MIP1-alpha; MIP-1-alpha(4-69); PAT 464.1; RP23-320E6.7; Scya3; SIS-alpha; SIS-beta; small inducible cytokine A3; small inducible cytokine A3 (homologous to mouse Mip-1 A); small-inducible cytokine A3; Tonsillar lymphocyte LD78 alpha protein; TY-5
Common Name CCL3 (MIP-1 alpha)
Gene Symbol Ccl3
Biological Activity Determined by its ability to chemoattract murine balb/c splenocytes using a concentration range of 10.0-100.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence APYGADTPTA CCFSYSRKIP RQFIVDYFET SSLCSQPGVI FLTKRNRQIC ADSKETWVQE YITDLELNA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.