Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse CCL4/MIP-1 beta Quantikine ELISA Kit

Catalog No. MMB00
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMB00 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MMB00 Supplier R&D Systems™ Supplier No. MMB00
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse CCL4/MIP-1 beta Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse CCL4/MIP-1 beta Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse CCL4/MIP-1 beta in cell culture supernates (50 ul), serum (25 ul), edta plasma (25 ul).

Specifications

Assay Range 7.80 - 500pg/mL
Assay Sensitivity 3pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA), Serum
Specificity Natural and Recombinant Mouse MIP-1 beta
For Use With (Application) ELISA
Gene ID (Entrez) 20303
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Serum (25uL),EDTA Plasma (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, Exodus-3, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP1 beta, MIP-1 beta, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1 b), Small-inducible cytokine A4, T-cell activation protein 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.