Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse CCL5 (RANTES) Recombinant Protein, PeproTech®

Catalog No. 25007100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25007100UG 100 μg
250-07-1MG 1 mg
250-07-20UG 20 μg
25007250UG 250 μg
25007500UG 500 μg
250-07-50UG 50 μg
250-07-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 25007100UG Supplier Gibco™ Supplier No. 25007100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine RANTES is a 7.8 kDa protein containing 68 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CCL5 (RANTES) is a chemokine gene located on the q-arm of chromosome 17. Chemokines are a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. CCL5, a member of the CC subfamily, functions as a chemoattractant for blood monocytes, memory T helper cells, and eosinophils. It causes the release of histamine from basophils and activates eosinophils. This cytokine is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor chemokine (C-C motif) receptor 5 (CCR5), and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor. Alternative splicing results in multiple transcript variants that encode different isoforms. Diseases associated with CCL5 include Human Immunodeficiency Virus Type 1 and Periapical Granuloma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P30882
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 20304
Molecular Weight (g/mol) 7.8 kDa
Name Mouse CCL5 (RANTES)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias beta-chemokine RANTES; CC chemokine; C-C motif chemokine 5; C-C motif chemokine ligand 5; CCL5; chemokine (C-C motif) ligand 5; D17S136E; EoCP; eosinophil chemotactic cytokine; MGC17164; MuRantes; OTTHUMP00000197106; RANTES; RANTES protein; RANTES(3-68); RANTES(4-68); regulated upon activation normal T-cell expressed and secreted; regulated upon activation, normally T-expressed, and presumably secreted; Scya5; SIS d; SISd; SIS-delta; SISd; small inducible cytokine A5; small inducible cytokine subfamily A (Cys-Cys), member 5; small-inducible cytokine A5; T cell-specific protein P228; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228; Unknown (protein for MGC:127014)
Common Name CCL5 (RANTES)
Gene Symbol CCL5
Biological Activity Determined by its ability to chemoattract human activated T cells cultured in the presence of IL-2 using a concentration range of 10-30 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMS
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.