Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse CCL7 (MCP-3) Recombinant Protein, PeproTech®

Catalog No. 2500850UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
250-08-50UG 50 μg
25008100UG 100 μg
250-08-10UG 10 μg
250-08-1MG 1 mg
25008250UG 250 μg
250-08-2UG 2 μg
25008500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 250-08-50UG Supplier Gibco™ Supplier No. 2500850UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine MCP-3 is an 8.5 kDa protein containing 74 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This is a secreted chemokine which attracts macrophages during inflammation and metastasis. It is a member of the C-C subfamily of chemokines which are characterized by having two adjacent cysteine residues. The protein is an in vivo substrate of matrix metalloproteinase 2, an enzyme which degrades components of the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q03366
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 20306
Molecular Weight (g/mol) 8.5 kDa
Name Mouse CCL7 (MCP-3)
Quantity 50 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C-C motif chemokine 7; C-C motif chemokine ligand 7; CCL7; chemokine (C-C motif) ligand 7; chemotactic protein-3; Fic; H-MCP-3; Intercrine/chemokine MARC; marc; Mcp3; MCP-3; MGC138463; MGC138465; Monocyte chemoattractant protein 3; monocyte chemotactic protein 3; NC28; Protein FIC; RANTES/sis homolog; RP23-350G1.4; SCYA6; Scya7; small inducible cytokine A7; small inducible cytokine A7 (monocyte chemotactic protein 3); small-inducible cytokine A7
Common Name CCL7 (MCP3)
Gene Symbol CCL7
Biological Activity Determined by its ability to chemoattract BaF3 cells expressing human CCR2 receptor using a concentration range of 300-500 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence QPDGPNASTC CYVKKQKIPK RNLKSYRRIT SSRCPWEAVI FKTKKGMEVC AEAHQKWVEE AIAYLDMKTP TPKP
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.