Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCR1 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-4712387
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5986A025 25 Tests
FAB5986A100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB5986A025 Supplier R&D Systems Supplier No. FAB5986A025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

CCR1 Monoclonal specifically detects CCR1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR1
Applications Flow Cytometry
Classification Monoclonal
Clone 643854
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. P51675
Gene Alias C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R
Gene Symbols CCR1
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCR1 Extracellular regions (aa 1-34, aa 92-107, aa 172-192, aa 265-281) Accession # P51675
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Primary or Secondary Primary
Gene ID (Entrez) 1230
Test Specificity Detects mouse CCR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CCR4, 5, or 8 is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.