Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD133 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7146568
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11332SP 25 μg
MB11332100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB11332SP Supplier R&D Systems Supplier No. MAB11332SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD133 Monoclonal specifically detects CD133 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen CD133
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 217106
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 5-25 μg;/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. O54990
Gene Alias AC133, AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, PROM1, prominin (mouse)-like 1, prominin 1, PROML1
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse CD133, Accession No. O54990
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 8842.0
Test Specificity Detects mouse CD133 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.