Learn More
Specifications
Specifications
| Description | Enrichment Kit |
| Target | Mouse CD3+ T Cell, Large Column |
| For Use With (Application) | Flow Cytometry |
| Product Type | Gene Symbol: CD3 |
| Content And Storage | Store at 4°C. Do not freeze. |
| Product Line | Novus |
| Quantity | 1 Column |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
