Learn More
Description
- Also known as: CLM4; DIgR1; Igsf7; Lmir2; Cd300c; Cd300d; MAIR-II
- LMIR2 is a type I transmembrane glycoprotein that belongs to the immunoregulatory signaling (IRS) family of molecules within the immunoglobulin (Ig) superfamily.
- LMIR2 is present on the surface of natural killer cells, granulocytes, most myeloid cells, dendritic cells, and a subpopulation of T and B lymphocytes.
- Mouse LMIR2 has the characteristics of an activatory molecule capable of inducing cellular activation and effector function in most cells and macrophages.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | CD300c2 |
| Accession Number | NP_598919 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 700 bp |
| Gene ID (Entrez) | 10871 |
| Gene Alias | CD300c antigenCMRF35 antigen, CD300c molecule, CLM4, CLM-6, CMRF-35, CMRF35 leukocyte immunoglobulin-like receptor, CMRF-35 A, CMRF35A leukocyte immunoglobulin-like receptor, CMRF35ACMRF35A1, CMRF35CMRF35-A1, CMRF35-like molecule 6, IgSF16, IGSF16CD300 antigen-like family member C, IGSF7, Immunoglobulin superfamily member 16, LIR, LMIR2, MAIR-II |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
