Learn More
ACROBiosystems Mouse CD36 Protein Fc Tag
Supplier:  ACROBiosystems CD6M5254100UG
Encompass_Preferred
Mouse CD36 Protein Fc Tag (CD6-M5254) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Lys 439 (Accession # Q08857-1).
Catalog No. 50-253-5359
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.