Learn More
ACROBiosystems ACROBiosystems Mouse CD39 / ENTPD1 Protein, His Tag
Supplier:  ACROBiosystems CD9M52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse CD39, His Tag (CD9-M52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 38 -Ile 478 (Accession # P55772-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 51.7 kDa.
Catalog No. 50-210-6414
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.