Learn More
Sino Biological Mouse CD3E Protein 1MG (Sino Biological)
Supplier:  Sino Biological 50410M02S1
Encompass_Preferred
A DNA sequence encoding the Mouse CD3E (NP_031674.1) (Met1-Asp108) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1193