Learn More
Sino Biological Mouse CD4 PAb 100UL 50134-T16-100
Supplier:  Sino Biological 50134T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD4 (Catalog#50134-M08H; NP_038516.1; Met1-Thr394). CD4 specific IgG was purified by Mouse CD4 affinity chromatography.
Catalog No. 50-241-2408
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.