Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD45 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB114500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB114500 500 μg
MAB114SP 25 μg
MAB114100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB114500 Supplier R&D Systems Supplier No. MAB114500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 8 publications

CD45 Monoclonal specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Complement-dependent Cytotoxicity, Immunocytochemistry, CyTOF-reported.

Specifications

Antigen CD45
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Cytotoxicity Assay, Immunocytochemistry
Classification Monoclonal
Clone 30-F11
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Immunoprecipitation, Complement-dependent Cytotoxicity, Immunocytochemistry 3-25 ug/mL, CyTOF-reported
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Gene Symbols PTPRC
Host Species Rat
Immunogen Mouse thymus and spleen cells
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5788
Test Specificity Detects mouse CD45. Recognizes all CD45 isoforms as well as the known mouse CD45 alloantigens (CD45-1/Ly 5a and CD45-2/Ly 5b) (15).
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.