Learn More
Sino Biological Mouse CD47 MAb 100UL 57231-R012-100
Supplier:  Sino Biological 57231R012100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD47 (rM CD47; Catalog#57231-M08H; Q61735-1; Met1-Lys140).
Catalog No. 50-240-7709
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.