Learn More
Sino Biological Mouse CD47 PAb 100UL 57231-T52-100
Supplier:  Sino Biological 57231T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD47 (rM CD47; Catalog#57231-M08H; Q61735-1; Met1-Lys140). CD47 specific IgG was purified by Mouse CD47 affinity chromatography.
Catalog No. 50-241-0266
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.