Learn More
Biomatik Corporation Mouse CD5 Antigen Like Protein (CD5L) ELISA Kit
Supplier:  Biomatik Corporation EKU03061
Encompass_Preferred
Mouse CD5 Antigen Like Protein (CD5L) ELISA Kit for detecting CD5L in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.216ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QWK4. Gene ID: 11801. Target Synonyms: API6; CT-2; SP-ALPHA; Spalpha; Apoptosis Inhibitor 6; Scavenger Receptor Cysteine Rich Family; IgM-associated peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4572
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.