Learn More
Description
- CD7 is a transmembrane protein which is a member of the immunoglobulin superfamily
- It is found on thymocytes and mature Tcells and plays an essential role in Tcell interactions and also in T-cell/Bcell interaction during early lymphoid development
- CD7 is a negative prognostic marker in myeloid malignancies
- The CD7 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mCd7 |
| Accession Number | NP_033984 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 646 bp |
| Gene ID (Entrez) | 924 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
