Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD8 alpha Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM116
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM116 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM116 Supplier R&D Systems Supplier No. BAM116
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

CD8 Monoclonal specifically detects CD8 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen CD8
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 53-6.7
Concentration LYOPH
Conjugate Biotin
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Alias CD8, CD8A, CD8a molecule, LEU2, p32
Gene Symbols CD8A
Host Species Rat
Immunogen Mouse thymus and spleen cells
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 925
Test Specificity Detects mouse CD8 alpha . Reacts with alpha chains of mouse CD8 (Bierer, B.E. et al. (1989) Annu. Rev. Immunol. 7:579).
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.