Learn More
Abcam Mouse CD86 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB3150521X96TESTS
Encompass_Preferred
Mouse CD86 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse CD86 with a sensitivity of 2.137 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-8952
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.