Learn More
Sino Biological Mouse CFI Protein 1MG 5A1431-M08H-1
Supplier:  Sino Biological 5A1431M08H1
Encompass_Preferred
A DNA sequence encoding the Mouse CFI (Q61129) (Met1-Val603) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-241-2142
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.