Learn More
Description
- Also known as: 5930406N14Rik
- CLEC-1 is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily.
- Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cellcell signaling, glycoprotein turnover, and roles in inflammation and immune response.
- CLEC-1 may play a role in regulating dendritic cell function.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mClec1a |
| Accession Number | NP_780735 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 823 bp |
| Gene ID (Entrez) | 51267 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
