Learn More
R&D Systems™ Mouse CLEC-2/CLEC1B (NP_064369) VersaClone cDNA
Description
- Also known as: Clec2; Clec-2; 1810061I13Rik
- CLEC1B is a type II transmembrane glycoprotein and member of the Ctype lectin-like family of receptors
- It is expressed in myeloid cells and NK cells
- CLEC1B is the receptor for the platelet-aggregating snake venom protein rhodocytin and the molecule podoplanin, a transmembrane sialoglycoprotein that, when bound to CLEC1B, is involved in platelet aggregation, tumor metastasis, and lymphatic vessel formation
- The CLEC-2/CLEC-1B insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mClec1b |
| Accession Number | NP_064369 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 703 bp |
| Gene ID (Entrez) | 51266 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.