Learn More
Description
- Clec4d C-type lectin domain family 4, member d [ Mus musculus (house mouse) ]
- Also known as: mcl; Mpcl; Clecsf8
- CLEC4D is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily.
- Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signaling, glycoprotein turnover, and roles in inflammation and immune response
- CLEC4D is restricted to monocytes/ macrophages and serves as an endocytic receptor
- The CLEC4D/CLECSF8 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mClec4d |
| Accession Number | NP_034949 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 673 bp |
| Gene ID (Entrez) | 338339 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
