Learn More
Description
- Also known as: Kclr; D18063; Clecsf13; KUCR_MOUSE
- CLEC4F is a type II transmembrane glycoprotein member of the C-type lectin superfamily.
- CLEC4F is expressed on Kupffer cells in the liver, but not on macrophages in other tissues.
- CLEC4F preferentially binds galactose and N-acetylgalactosamine in a calciumdependent manner.
- Its activity at neutral, but not at acidic pH, suggests a capacity to internalize and release ligands into the endosomal system.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mClec4f |
| Accession Number | NP_058031 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1660 bp |
| Gene ID (Entrez) | 165530 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
