Learn More
ACROBiosystems ACROBiosystems Mouse Coagulation Factor III / Tissue Factor Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems TF3M52H3500UG
Encompass_Preferred
Recombinant Protein;500UG;Mouse Tissue Factor, His Tag (TF3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Glu 251 (Accession # P20352-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.3 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6527
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.