Learn More
American Research Products Inc Mouse coagulation factor VII
Supplier:  American Research Products Inc CSBE17817M
Encompass
Mouse coagulation factor VII FVII ELISA Kit from CUSABIO Detection Range 0 45 ng/mL-30 ng/mL Sensitivity 0 11 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27033
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.