Learn More
American Research Products Inc Mouse Coagulation Factor XIII
Supplier: American Research Products Inc CSBE13519M
Mouse Coagulation Factor XIII A1 Polypeptide (F13A1) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich