Learn More
R&D Systems™ Mouse CX3CL1/Fractalkine Quantikine ELISA Kit
Description
Specifications
Specifications
| Assay Range | 0.62 - 40ng/mL |
| Assay Sensitivity | 0.32ng/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates |
| Specificity | Natural and Recombinant Mouse Fractalkine (chemokine and extracellular domains) |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 20312 |
| Quantity | 1 x 96-well Plate |
| Sample Volume | Cell Culture Supernates (50uL),Tissue Homogenates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.