Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse CXCL12/SDF-1 alpha Quantikine ELISA Kit
SDP

Catalog No. MCX120 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MCX120 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MCX120 Supplier R&D Systems™ Supplier No. MCX120
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse CXCL12/SDF-1 alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse CXCL12/SDF-1 alpha Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse CXCL12/SDF-1 alpha in cell culture supernates (10 ul), serum (50 ul).

Specifications

Assay Range 0.16 - 10ng/mL
Assay Sensitivity 0.069ng/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Serum
Specificity Natural and Recombinant Mouse SDF-1 alpha
For Use With (Application) ELISA
Gene ID (Entrez) 20315
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (10uL),Serum (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CXCL12 chemokine (C-X-C motif) ligand 12, IRH, PBSF, SCYB12, SDF1, SDF-1, SDF1 alpha, SDF1a, SDF1B, TLSF, TPAR1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.