Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse CXCL12 (SDF-1a) Recombinant Protein, PeproTech®

Catalog No. 25020A500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25020A500UG 500 μg
25020A100UG 100 μg
25020A10UG 10 μg
250-20A-1MG 1 mg
25020A250UG 250 μg
250-20A-2UG 2 μg
25020A50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 25020A500UG Supplier Gibco™ Supplier No. 25020A500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine SDF-1alpha is a 7.9 kDa protein containing 68 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P40224
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 20315
Molecular Weight (g/mol) 7.9 kDa
Name Mouse CXCL12 (SDF-1 A)
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); CXC; CXC motif chemokine; C-X-C motif chemokine 12; C-X-C motif chemokine ligand 12; CXCL; Cxcl12; H-SDF-1-alpha; Intercrine reduced in hepatomas; IRH; Pbsf; pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; SCYB12; SDF 1 A; SDF 1 alpha; SDF 1 A; SDF1; SDF-1; SDF1 alpha; SDF-1 gamma; SDF1A; SDF-1-alpha(3-67); SDF1B; SDF-1-beta(3-72); SDF1a; Stromal cell-derived factor 1; stromal cell-derived factor-1 alpha; stromal cell-derived factor-1 gamma; thymic lymphoma cell-stimulating factor; TLSF; Tpar1
Common Name CXCL12 alpha
Gene Symbol CXCL12
Biological Activity Determined by its ability to chemoattract human peripheral blood monocytes using a concentration of 50-100 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.