Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse CXCL13 (BLC/BCA-1) Recombinant Protein, PeproTech®

Catalog No. 250241MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
250-24-1MG 1 mg
25024100UG 100 μg
250-24-20UG 20 μg
25024250UG 250 μg
25024500UG 500 μg
250-24-50UG 50 μg
250-24-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 250-24-1MG Supplier Gibco™ Supplier No. 250241MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine BLC is a 9.8 kDa protein containing 88 amino acid residues including the four highly conserved cysteine residues present in CXC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CXCL13, also known as BCA-1 and Angie, is a small chemokine strongly expressed in the secondary lymphoid organs. It preferentially promotes the migration of B lymphocytes (both B-1 and B-2) to the follicles of the spleen, lymph nodes, and Peyer's patches. It is produced by macrophages, and in mouse the highest constitutive producers include large peritoneal macrophages, Kuppfer cells and lung macrophages (both CD11c+ and CD11c-). The cellular ligand for CXCL13 is CXCR5 also known as BLR-1 (Burkitt’s Lymphoma Receptor 1). CXCR5 is expressed on B cells, Tfh cells and, to a lower degree, some memory T cell populations. CXCL13 plays an important role in the development of chronic lymphocytic leukemia (CCL). CXCL13 is also a plasma biomarker for germinal center activity and generation of broadly neutralizing antibodies against HIV.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O55038
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 55985
Molecular Weight (g/mol) 9.8 kDa
Name Mouse CXCL13 (BLC/BCA-1)
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 4631412M08Rik; Angie; ANGIE2; B cell-attracting chemokine 1; b lymphocyte chemoattractant; BCA1; BCA-1; B-cell chemoattractant; B-cell-attracting chemokine 1; B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1); BLC; BLR1L; B-lymphocyte chemoattractant; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chem; chemokine (C-X-C motif) ligand 13; chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant); CXC; CXC chemokine BLC; CXC motif chemokine; CXC motif chemokine 13; C-X-C motif chemokine 13; C-X-C motif chemokine ligand 13; CXCL; Cxcl13; SCYB13; small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cell chemoattractant); small inducible cytokine subfamily B (Cys-X-Cys), member 13; small-inducible cytokine B13
Common Name CXCL13
Gene Symbol Cxcl13
Biological Activity Determined by its ability to chemoattract human B cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence ILEAHYTNLK CRCSGVISTV VGLNIIDRIQ VTPPGNGCPK TEVVIWTKMK KVICVNPRAK WLQRLLRHVQ SKSLSSTPQA PVSKRRAA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.