Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse CXCL2 (GRO beta) Recombinant Protein, PeproTech®

Catalog No. 25015250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25015250UG 250 μg
25015100UG 100 μg
250-15-1MG 1 mg
250-15-20UG 20 μg
25015500UG 500 μg
250-15-50UG 50 μg
250-15-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 25015250UG Supplier Gibco™ Supplier No. 25015250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine MIP-2 is a 7.8 kDa protein consisting of 73 amino acids, including the 'ELR' motif common to the CXC chemokine family that binds to CXCR1 or CXCR2. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10889
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 20310
Molecular Weight (g/mol) 7.8 kDa
Name Mouse CXCL2 (GRO beta)
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 2; CINC-2 A; Cinc3; CINC-3; CXC; CXC motif chemokine; C-X-C motif chemokine 2; C-X-C motif chemokine ligand 2; CXCL; CXCL2; Cytokine-induced neutrophil chemoattractant 3; GRO b; gro beta; GRO β; GRO2; GRO2 oncogene; GROb; GRObeta; gro-beta; GRO-beta(5-73); GRO-beta-T; Growth-regulated protein beta; GROβ; Hematopoietic synergistic factor; HSF; Macrophage inflammatory protein 2; macrophage inflammatory protein 2-alpha; melanoma growth stimulatory activity beta; MGSA beta; Mgsa-b; MIP 2 A; MIP 2 alpha; MIP 2 A; Mip2; MIP-2; MIP2 a; MIP2 a; MIP2A; MIP-2 A; MIP2-alpha; SB-251353; Scyb; Scyb2; small inducible cytokine subfamily, member 2
Common Name CXCL2
Gene Symbol Cxcl2
Biological Activity Determined by its ability to chemoattract total human neutrophils using a concentration range of 1.0-10.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence AVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KAN
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.