Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse CXCL2/MIP-2 Quantikine ELISA Kit

Catalog No. MM200
Change view
Click to view available options
Quantity:
2 Plates
6 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MM200 2 Plates
SMM200 6 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MM200 Supplier R&D Systems™ Supplier No. MM200
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse CXCL2/MIP-2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse CXCL2/MIP-2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse CXCL2/GRO beta/MIP-2/CINC-3 in cell culture supernates (50 ul), serum (50 ul).

Specifications

Assay Range 7.80 - 500pg/mL
Assay Sensitivity 1.5pg/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Serum
Specificity Natural and Recombinant Mouse MIP-2
Sufficient For 2 x 96-well Plates
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 20310
Quantity 2 Plates
Sample Volume Cell Culture Supernates (50uL),Serum (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym chemokine (C-X-C motif) ligand 2, CINC-2 A, CINC3, CINC-3, C-X-C Motif Chemokine 2, GRO beta, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MGSA-beta, MIP2, MIP-2, MIP-2 A, MIP2A, MIP2-alpha, SCYB2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.