Learn More
Kingfisher Biotech Inc Mouse CXCL4 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1712M500
Encompass_Preferred
The Mouse CXCL4 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Mouse CXCL4 applications are for cell culture, ELISA standard, and Western Blot Control. Mouse CXCL4 yeast-derived recombinant protein can be purchased in multiple sizes. Mouse CXCL4 Specifications: (Molecular Weight: 8.2 kDa) (Amino Acid Sequence: VTSAGPEESD GDLSCVCVKT ISSGIHLKHI TSLEVIKAGR HCAVPQLIAT LKNGRKICLD RQAPLYKKVI KKILES (76)) (Gene ID: 56744). For research use only. Made in the USA
Catalog No. 50-253-2905
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.