Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CXCL4/PF4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB595100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB595100 100 μg
MAB595SP 25 μg
MAB595500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB595100 Supplier R&D Systems Supplier No. MAB595100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 4 publications

CXCL4/PF4 Monoclonal specifically detects CXCL4/PF4 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CXCL4/PF4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 140910
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Z126
Gene Alias chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF4, PF-4, platelet factor 4, SCYB4oncostatin-A
Gene Symbols PF4
Host Species Rat
Immunogen E. coli-derived recombinant mouse CXCL4/PF4 Val30-Ser105 Accession # Q9Z126
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5196
Test Specificity Detects mouse CXCL4/PF4 in ELISAs and Western blots. In ELISAs, this antibody does not cross-react or interfere recombinant human (rh) CXCL/PF4, recombinant mouse (rm) CXCL9/MIG, rmGM-CSF, rmCXCL11/I-TAC, rhCXCL8/IL-8, rmIL-5, or rhCXCL10/IP-10. In Western blots, no cross-reactivity was observed with rhGCP-2, rmGCP-2, rhGRO alpha , rhGRO beta , rhGRO gamma , rhMIP-1 alpha , rmMIP-1 alpha , rmMIP-1 beta , rhMIP-1 delta , rmMIP-1 gamma , rmMIP-2, rhMIP-3 alpha , rmMIP-3 alpha , rrMIP-3 alpha , rhMIP-3 beta , or rmMIP-3 beta ,
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.