Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse CXCL9/MIG Quantikine ELISA Kit
SDP

Catalog No. MCX900 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MCX900 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MCX900 Supplier R&D Systems™ Supplier No. MCX900

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse CXCL9/MIG Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse CXCL9/MIG Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse CXCL9/MIG in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (25 ul), edta plasma (25 ul).

Specifications

Assay Range 31.20 - 2000pg/mL
Assay Sensitivity 7.8pg/mL
Conjugate HRP
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Plasma (EDTA), Serum, Tissue Homogenates
Specificity Natural and Recombinant Mouse MIG
For Use With (Application) ELISA
Gene ID (Entrez) 17329
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Tissue Homogenates (50uL),Serum (25uL),EDTA Plasma (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIG, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.