Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Cyclophilin B Recombinant Protein
SDP

Catalog No. p-7112723 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB59665100U 100 μg
NB5966520UG 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB59665100U Supplier Novus Biologicals™ Supplier No. NBP259665100UG
Only null left
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 34-216 of Mouse Cyclophilin B The Recombinant Mouse Cyclophilin B Protein is derived from E. coli. The Recombinant Mouse Cyclophilin B Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

For Use With (Application) PAGE
Formulation In PBS (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 5479
Molecular Weight (g/mol) 22.7 kDa
Name Ppib Recombinant Protein Antigen
Purification Method >95% by SDS-PAGE
Quantity 100 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias Cyclophilin B, cyclophilin-like protein
Gene Symbol Ppib
Product Type Recombinant Protein Antigen
Conjugate Unconjugated
Cross Reactivity Mouse
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.