Learn More
Description
- Clec4a2 C-type lectin domain family 4, member a2 [ Mus musculus (house mouse) ]
- Also known as: DCIR; Dcir1; Clec4a; Clecsf6
- DCIR, also known as lectin-like immunoreceptor (LLIR), is a type II transmembrane protein belonging to the C-type lectin domain family and is designated CLEC4A (previously designated CLECSF6).
- Four transcript variants encoding distinct isoforms have been identified.
- DCIR contains one carbohydrate recognition domain in its C-terminal extracellular domain and an immunoreceptor tyrosine-based inhibitory motif (ITIM) in its cytoplasmic domain.
- DCIR is expressed on dendritic cells, B cells, monocytes/macrophages and granulocytes.
- The DCIR/CLEC4A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mClec4a2 |
| Accession Number | NP_036129 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 730 bp |
| Gene ID (Entrez) | 50856 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
