Learn More
Sino Biological Mouse DDR2 PAb 100UL 50541-RP02-100
Supplier:  Sino Biological 50541RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse DDR2 (rM DDR2; Catalog#50541-M08H; NP_072075.2; Met 1-Arg 399). DDR2 specific IgG was purified by Mouse DDR2 affinity chromatography.
Catalog No. 50-241-0534
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.